ML-9 ≥98% (by TLC)
50 mg
ENZOBMLEI1530050
Selective inhibitor of MLC kinase (Ki=3.8 µM). Inhibits insulin-stimulated 2-deoxyglucose transport and PP-1 activation in adipocytes, possibly via an effect on MAP kinase or its activation. Useful PKB inhibitor (IC50=10-50 µM). Also known to inhibit PKA (IC50=20 µM) and S6 kinase (IC50=50 µM). Does not inhibit PI 3-kinase.
- Myelin basic protein inhibitor
White solid.
Soluble in DMSO (10mg/ml) and ethanol/water (1:1, 10mg/ml).
: 1-(5-Chloronaphthalene-1-sulphonyl)-1H-hexahydro-1,4-diazepine hydrochloride, 1-((5-Chloronaphthalen-1-yl)sulphonyl)-1,4-diazepane hydrochloride
- :105637-50-1
- :C₁₅H₁₇N₂O₂SCl·HCl
- :MFCD00065525
- :361.3 g/mol
- :Refrigerator
- Pk:50 mg