Order Entry
Poland
ContactUsLinkComponent
 
Chaetocin ≥98% (by TLC)
  ENZOBMLGR3490200
 :  
Chaetocin ≥98% (by TLC)
  ENZOBMLGR3490200
 :  

 

  • Pk:
    200 µG
  • Seq:
    1358

 

 

 :
 :696.839 g/mol
 :28097-03-2
 :C₃₀H₂₈N₆O₆S₄
 :MFCD00133163