Simvastatin sodium salt ≥97% (by HPLC)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:101314-97-0
:
:458.6 g/mol
Frequently Bought Together
VWR®, Strips of PCR Tubes and Caps
Supplier: VWR Collection
VWR®, Test Tubes without Rim
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Cryoboxes, 133×133 mm
Supplier: RATIOLAB
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
VWR®, Cryo coder cap inserts
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Nunc-Immuno™ modules
Supplier: Thermo Fisher Scientific
Round-bottom polystyrene tubes, disposable, Falcon®
Supplier: Corning
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
J.T.Baker®, Plates, Square Well
Supplier: Avantor
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
Microplates, 96-well
Supplier: Greiner Bio-One
96 MicroWell™ plates, Nunclon™∆
Supplier: Thermo Fisher Scientific
Sampling spoon, large, Sterileware®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP