N-Palmitoyl phosphatidylethanolamine ≥98% (by TLC), white solid
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:108321-22-8
:
Formula:C₅₃H₁₀₄NO₉P
:930.4 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity:≥98% (TLC)
Specification Test Results
Frequently Bought Together