Order Entry
Poland
ContactUsLinkComponent
 
Flammable
Irritant
Health
12(S)-Hydroxy-(5Z,8Z,10E,14Z)-eicosatetraenoic acid 50 µg/ml in ethanol ≥98% (by HPLC)
Catalog # ENZOBMLH0120050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Flammable
Irritant
Health
12(S)-Hydroxy-(5Z,8Z,10E,14Z)-eicosatetraenoic acid 50 µg/ml in ethanol ≥98% (by HPLC)
Catalog # ENZOBMLH0120050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Seq:
    1202

Specifications

Specification Test Results

  • Concentration:
    50 µg/ml
  • Formulation:
    Solution in ethanol
  • Purity:
    ≥98% (HPLC)
  • Identity:
    Determined by NMR

Specification Test Results

About this item

 :
 :320.47 g/mol
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
UN:1170
ADR:3,III
 :GHS02
 :GHS07
 :GHS08
CAS Number:54397-83-0
Formula:C₂₀H₃₂O₃
MDL Number:MFCD00036916