Order Entry
Poland
ContactUsLinkComponent
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) ≥98% (by HPLC), Ultrapure
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) ≥98% (by HPLC), Ultrapure
Catalog # ENZOENZ52156
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) ≥98% (by HPLC), Ultrapure
Catalog # ENZOENZ52156
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :326.19 g/mol
Storage Temperature:Freezer
CAS Number:162558-52-3
Formula:C₁₄H₁₆NO₃Br
MDL Number:MFCD00467854