Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
alpha-Hydroxytriazolam
α-Hydroxytriazolam ≥96% (by TLC)
α-Hydroxytriazolam ≥96% (by TLC)
α-Hydroxytriazolam ≥96% (by TLC)
ENZOBMLJ1300001
:
Enzo Life Sciences
:
α-Hydroxytriazolam ≥96% (by TLC)
ENZOBMLJ1300001
:
Enzo Life Sciences
:
Pk:
1 mg
Seq:
2070
Purity:
≥96% (TLC)
Identity:
Determined by H-NMR and MS
:
:
359.2 g/mol
:
GHS07
:
GHS08
:
Freezer
:
37115-45-0
:
C₁₇H₁₂Cl₂N₄O