Order Entry
Poland
ContactUsLinkComponent
α-Hydroxyalprazolam ≥98% (by TLC)
Irritant
α-Hydroxyalprazolam ≥98% (by TLC)
Catalog # ENZOBMLJ1250005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Irritant
α-Hydroxyalprazolam ≥98% (by TLC)
Catalog # ENZOBMLJ1250005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Purity:
    ≥98% (TLC)

Specification Test Results

About this item

 :
 :324.8 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:37115-43-8
Formula:C₁₇H₁₃CIN₄O