
SCH-202676 ≥99% (by HPLC), yellow solid
Catalog # ENZOBMLG5210010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
Specification Test Results
- Purity:≥99% (HPLC)
- Identity:Determined by NMR
Specification Test Results
About this item
:
:348.3 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:70375-43-8
Formula:C₁₅H₁₃N₃S·HBr