0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
Anandamide (18:2, n-6)
Anandamide (18:2, n-6) ≥98% (by TLC)
Anandamide (18:2, n-6) ≥98% (by TLC)
ENZOBMLFA0260005
:
ENZO LIFE SCIENCES
:
Anandamide (18:2, n-6) ≥98% (by TLC)
ENZOBMLFA0260005
:
ENZO LIFE SCIENCES
:
Pk:
5 mg
Seq:
2210
Purity:
≥98% (TLC)
:
:
323.5 g/mol
:
Ultra-Low Temperature Freezer (–80°C)
:
68171-52-8
:
C₂₀H₃₇NO₂