Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tyrphostin B7
Tyrphostin B7 ≥99% (by TLC)
Tyrphostin B7 ≥99% (by TLC)
:
Enzo Life Sciences
Tyrphostin B7 ≥99% (by TLC)
:
Enzo Life Sciences
:
134036-53-6
:
:
C₁₅H₉N₅
:
MFCD00236449
:
300…302 °C
:
287.3 g/mol
:
Freezer
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
: Corning
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
: VWR Collection