Order Entry
Poland
ContactUsLinkComponent
MIF antagonist, ISO-1 ≥97% (by HPLC)
MIF antagonist, ISO-1 ≥97% (by HPLC)
  ENZOENZCHM1210005
 :  
undefined
MIF antagonist, ISO-1 ≥97% (by HPLC)
  ENZOENZCHM1210005
 :  

 

  • Pk:
    5 mg
  • Seq:
    2210

 

 

 :
 :235.2 g/mol
 :478336-92-4
 :C₁₂H₁₃NO₄
 :MFCD22376813