Order Entry
Poland
ContactUsLinkComponent
Dihydrorhodamine 123 ≥95% (by HPLC), Ultrapure
Dihydrorhodamine 123 ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52302
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
undefined
Dihydrorhodamine 123 ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52302
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Methyl 2-(3,6-diamino-9H-xanthen-9-yl)benzoate, 2-(3,6-Diamino-9H-xanthene-9-yl)-benzoic acid methyl ester
 :
 :346.386 g/mol
Storage Temperature:Freezer
CAS Number:109244-58-8
Formula:C₂₁H₁₈N₂O₃
MDL Number:MFCD04221428