0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
Isotetrandrine
Isotetrandrine ≥98% (by TLC)
Isotetrandrine ≥98% (by TLC)
Isotetrandrine ≥98% (by TLC)
Catalog # ENZOBMLG5200001
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
Isotetrandrine ≥98% (by TLC)
Catalog # ENZOBMLG5200001
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
622.762 g/mol
CAS Number:
477-57-6
Formula:
C₃₈H₄₂N₂O₆
MDL Number:
MFCD00236404
Product Details