Order Entry
Poland
ContactUsLinkComponent
Melengestrol acetate ≥97% (by HPLC)
Toxic
Irritant
Health
Environment
Melengestrol acetate ≥97% (by HPLC)
Catalog # ENZOBMLS5200200
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Toxic
Irritant
Health
Environment
Melengestrol acetate ≥97% (by HPLC)
Catalog # ENZOBMLS5200200
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Purity:
    ≥97% (HPLC)
  • Identity:
    Determined by IR and UV spectra.

Specification Test Results

About this item

 :
 :396.527 g/mol
UN:3077
ADR:9,III
 :GHS06
 :GHS07
 :GHS08
 :GHS09
Storage Temperature:Ambient
CAS Number:2919-66-6
Formula:C₂₅H₃₂O₄