Order Entry
Poland
ContactUsLinkComponent
(-)-Bicuculline methiodide (synthetic) ≥98% (by HPLC)
Toxic
Environment
(-)-Bicuculline methiodide (synthetic) ≥98% (by HPLC)
Catalog # ENZOBMLEA1490050
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Toxic
Environment
(-)-Bicuculline methiodide (synthetic) ≥98% (by HPLC)
Catalog # ENZOBMLEA1490050
Supplier:  Enzo Life Sciences
CAS Number:  

Specification Test Results

  • Source:
    Synthetic
  • Purity:
    ≥98% (HPLC)

Specification Test Results

About this item