Order Entry
Poland
ContactUsLinkComponent
Palmitoylethanolamide ≥98% (by TLC)
Palmitoylethanolamide ≥98% (by TLC)
Catalog # ENZOBMLFA0180050
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Palmitoylethanolamide ≥98% (by TLC)
Catalog # ENZOBMLFA0180050
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :299.499 g/mol
Storage Temperature:Refrigerator
CAS Number:544-31-0
Formula:C₁₈H₃₇NO₂
MDL Number:MFCD00020562