Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dihydroceramide C6
Dihydroceramide C6 ≥98%
Dihydroceramide C6 ≥98%
ENZOBMLSL1110005
:
ENZO LIFE SCIENCES
:
Dihydroceramide C6 ≥98%
ENZOBMLSL1110005
:
ENZO LIFE SCIENCES
:
Pk:
5 mg
Seq:
2210
Purity:
≥98%
:
:
399.66 g/mol
:
Freezer
:
171039-13-7
:
C₂₄H₄₉NO₃
:
MFCD01320384