Order Entry
Poland
ContactUsLinkComponent
WAY-200070 ≥98% (by TLC)
Toxic
Irritant
WAY-200070 ≥98% (by TLC)
Catalog # ENZOBMLGR2470010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Toxic
Irritant
WAY-200070 ≥98% (by TLC)
Catalog # ENZOBMLGR2470010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

Specification Test Results

  • Purity:
    ≥98% (TLC)
  • Appearance:
    Off-white solid

Specification Test Results

About this item

 :
 :306.115 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :GHS06
 :GHS07
CAS Number:440122-66-7
Formula:C₁₃H₈BrNO₃
MDL Number:MFCD16618385