Order Entry
Poland
ContactUsLinkComponent
Prostaglandin F2α tris salt (synthetic) ≥99% (by HPLC)
Prostaglandin F2α tris salt (synthetic) ≥99% (by HPLC)
Catalog # ENZOBMLPG0090010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Prostaglandin F2α tris salt (synthetic) ≥99% (by HPLC)
Catalog # ENZOBMLPG0090010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :475.623 g/mol
Storage Temperature:Freezer
CAS Number:38562-01-5
Formula:C₂₄H₄₅NO₈
MDL Number:MFCD00077863