Latanoprost ≥98%
5 mg
Catalog # ENZOBMLRA1200005
Supplier: Enzo Life Sciences
Jump to:
Product Details & Documents
Prostanoid receptor ligand.
Prodrug (isopropyl ester) of 17-phenyl-13,14-dihydro-prostaglandin F₂α. The free acid form is a potent agonist at FP receptors. It is in clinical use as a topical antiglaucoma agent.
Synonyms: Latanoprost, (Z)-Isopropyl 7-((1R,2R,3R,5S)-3,5-dihydroxy-2-((R)-3-hydroxy-5-phenylpentyl)cyclopentyl)hept-5-enoate
- CAS Number:130209-82-4
- Formula:C₂₆H₄₀O₅
- MDL Number:MFCD00216074
- Molecular Weight:432.60 g/mol
- Storage Temperature:Freezer
Specifications
Specification Test Results
- Purity:≥98% (Assay)
- Identity:Determined by NMR
Specifications
- Pk:5 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



