D-erythro-Sphingosine ≥98% (by TLC)
Catalog # ENZOBMLEI1550100
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:299.50 g/mol
Storage Temperature:Freezer
CAS Number:123-78-4
Formula:C₁₈H₃₇NO₂
MDL Number:MFCD00036751