Order Entry
Poland
ContactUsLinkComponent
 
5,6-Dichlorobenzimidazole-1-β-D-Ribofuranoside ≥97% (by TLC)
  ENZOBMLEI2310050
 :  
undefined
5,6-Dichlorobenzimidazole-1-β-D-Ribofuranoside ≥97% (by TLC)
  ENZOBMLEI2310050
 :  

 

  • Pk:
    50 mg
  • Seq:
    2370

 

 

 :
 :319.14 g/mol
 :Freezer
 :53-85-0
 :C₁₂H₁₂Cl₂N₂O₄
 :MFCD00036785