Order Entry
Poland
ContactUsLinkComponent
(S)-(+)-Ketoprofen ≥98.5% (by HPLC)
(S)-(+)-Ketoprofen ≥98.5% (by HPLC)
Catalog # ENZOBMLEI2880001
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
(S)-(+)-Ketoprofen ≥98.5% (by HPLC)
Catalog # ENZOBMLEI2880001
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :254.29 g/mol
Storage Temperature:Ambient
CAS Number:22161-81-5
Formula:C₁₆H₁₄O₃
MDL Number:MFCD00673316