Anti-TSWV Mouse Monoclonal [clone: 1C1.A3 HL1]
Catalog # AGRIAS204517
Supplier: Agrisera
Specifications
- Antigen name:Tomato Spotted Wilt Virus
- Antigen symbol:TSWV
- Clonality:Monoclonal
- Clone:1C1.A3 HL1
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Western blot:Yes
- Purity:Protein G purified immunoglobulin in PBS pH 7,4, 150 mM NaCl, 0,05% sodium azide
- Storage temperature:Store at −20 °C. Make aliquots to avoid repeated freeze-thaw cycles. Please, remember to spin tubes briefly prior to opening them to avoid any losses that might occur from lyophilized material adhering to the cap or sides of the tubes.
- Immunogen:Non-structural protein encoded by the small RNA of TSWV
- Tested applications:DAS-ELISA
- Pk:100 µG
Specifications
About this item
Tomato spotted wilt tospovirus (TSWV) is the type member of the plant-infecting viruses of the genus Tospovirus in the family Bunyaviridae. TSWV is transmitted by thrips (Thysanoptera: Thripidae) to over 650 different plant species in temperate and tropical regions.
- Recommended dilution - 1:500 - 1:1000 (ELISA), (WB)
Once the plants infected with the virus, they result in spotting, wilting, reduced vegetative output and death of the plant. Some of the crops known to be affected by the viruses include, peanuts, watermelons, tomatoes, zucchinis and others.
Type:
Antigen: TSWV
Clonality: Monoclonal
Clone: 1C1.A3 HL1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Tomato Spotted Wilt Virus (TSWV)