Anti-PS Mouse Monoclonal Antibody
Catalog # AGRIAS204489
Supplier: Agrisera
Specifications
- Antigen name:Phytochelatin Synthase
- Antigen symbol:PS
- Clonality:Monoclonal
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Western blot:Yes
- Purity:Protein G purified immunoglobulin in PBS pH 7,4, 150 mM NaCl, 0,05% sodium azide
- Molecular weight:57 kDa
- Storage temperature:Store at −20 °C. Make aliquots to avoid repeated freeze-thaw cycles. Please, remember to spin tubes briefly prior to opening them to avoid any losses that might occur from lyophilized material adhering to the cap or sides of the tubes.
- Immunogen:N-terminal peptide conjugated to MAP (Multiple antigenc peptide)
- Pk:100 µG
Specifications
About this item
Together with gamma - glutamyl Cys sythetase (γ-ECS) and glutathione synthetase, phytochelatin synthetase (PS), is an enzyme involved in, and required for, synthetizing phytochelatins (PC).
- Recommended dilution - 1:1000 (ELISA), (WB)
In certain plants, deficiency of such enzymes leads to low tolerance to metal contamination while mutants over-expressing them are highly tolerant to metal contamination and may be used in phyto-remediation of heavy metals.
Type:
Antigen: PS
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Schizosaccharomyces pombe