Order Entry
Poland
ContactUsLinkComponent
Mycoplasma removal agent, MP Biomedicals
Mycoplasma removal agent, MP Biomedicals
Catalog # ICNA093050044
Supplier:  MP Biomedicals
CAS Number:  
undefined
Mycoplasma removal agent, MP Biomedicals
Catalog # ICNA093050044
Supplier:  MP Biomedicals
CAS Number:  
Specifications
  • Pk:
    5 mL
  • Description:
    Mycoplasma removal agent
Specifications
About this item
Mycoplasma removal agent (MRA) is an effective solution for the problem of mycoplasma contamination in cell cultures.
  • Completely kills mycoplasma with no recurrence of contamination
  • Effective at low concentration
MRA is a derivative of the quinoline family of antibiotics. It eliminates mycoplasma infection (recommended use at a concentration of 0,5 μg/ml) by inhibiting mycoplasma DNA gyrase. Cell toxicity is low following treatment at the recommended concentration, as is regrowth of mycoplasma after cellular transfer.
Delivery: Supplied as a ready to use solution and each vial can decontaminate up to 25 cell cultures.