Order Entry
Poland
ContactUsLinkComponent
 
 
Nitric acid Reag. Ph. Eur. 1058402
  87880.180
 :  
undefined
 
Nitric acid Reag. Ph. Eur. 1058402
  87880.180
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

 

 

  • Performance test:
    passes test (125 g/l)

 

 

 :
 :63.01 g/mol
 :~86 °C (1013 hPa)
 :–42 °C
 :1.2…1.4 g/cm³ (20 °C)
 :Ambient
 :231-714-2
 :2031
 :8,II
 :GHS05
 :7697-37-2
 :HNO₃
 :MFCD00011349