Order Entry
Poland
ContactUsLinkComponent
L-Alanine, Sigma-Aldrich®
L-Alanine, Sigma-Aldrich®
Supplier:  Merck
Certificates
L-Alanine, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    166.5 g/l (25 °C)
  • Melting Point:
    295 - 297 °C (decomposition)
  • Molar Mass:
    89.09 g/mol
  • Bulk Density:
    660 kg/m3
  • Density:
    1.40 g/cm3 (20 °C)
  • pH:
    5.5 - 6.5 (100 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together