Order Entry
Poland
Orders LinkContactUsLinkComponent
Tris pH Buffer Solution

Tris pH Buffer Solution

rr tris-HCl buffer, 8.5, HCl, Clear, 1000 ml, 1 L
Catalog # CHMP177665014.1000
Supplier:   CHEMPUR
Tris pH Buffer Solution
Catalog # CHMP177665014.1000
Supplier:  CHEMPUR
CAS Number:  
Tris pH Buffer Solution

rr tris-HCl buffer, 8.5, HCl, Clear, 1000 ml, 1 L

CAS Number:  
Price
242 zł
Log in for availability
1 * 1 L
Product Details & Documents
  • Solubility in water: unlimited
  • Flash point: closed cup: none
Specifications

Specifications

  • pH buffer type:
    HCl
  • Pk:
    1 L
  • Colour:
    Clear
  • Description:
    rr tris-HCl buffer
  • Final volume:
    1000 ml
  • pH value:
    8.5
  • Cat. No.:
    CHMP177665014.1000
  • Odour/scent:
    Odourless
  • Physical form:
    Liquid
  • Concentration:
    0,1 mol
  • Density:
    approx. 1,00 g/cm³ (20˚C)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.