0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
Orders Link
ContactUsLinkComponent
Sodium hydroxide
Sodium hydroxide 30%
Sodium hydroxide 30%
Plastic can, 20 L
CHMP658109249.9020
:
CHEMPUR
Sodium hydroxide 30%
CHMP658109249.9020
:
CHEMPUR
:
1310-73-2
Sodium hydroxide 30%
Plastic can, 20 L
:
1310-73-2
813 zł
1 * 20 L
-
+
:
Caustic soda
:
1310-73-2
:
NaOH
:
MFCD00003548
:
40 g/mol
:
1824
Pack type:
Plastic can
Pk:
20 L
Hydrochloric acid 35 - 38%, pure
#: CHMP425752836.9010
: CHEMPUR