Order Entry
Poland
ContactUsLinkComponent
 
Corrosive
Sodium hydroxide 2.0 N in aqueous solution, Biotech Reagent, J.T.Baker®
Catalog # 0390-07
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Sodium hydroxide 2.0 N in aqueous solution, Biotech Reagent, J.T.Baker®
Catalog # 0390-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Jerrican, poly pail
  • Pk:
    19 L
  • Seq:
    5282

Specifications

Specification Test Results

  • GMP Manufactured Product:
    1.9 - 2.1
  • Protect from air to avoid absorption of carbon dioxide.:
    ≤5 ppm
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤1 ppm
  • Appearance (Clear, colorless liquid):
    Passes Test
  • Normality:
    1.9 - 2.1
  • Chloride (Cl):
    ≤ 5 ppm
  • ACS - Heavy Metals (as Pb):
    ≤ 1 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 0.5 ppm
  • Endotoxin Concentration (EU/mL:
    ≤ 2

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:109 °C (1013 hPa)
Density:≤1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,II
 :Filtered through a 0.2 µm filter.
EINECS:215-185-5
UN:1824
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548