L(-)-Sorbose 98%
:87-79-6
:
:C₆H₁₂O₆
:MFCD00151097
:180.16 g/mol
:12,08874
:380 °C (1013 hPa)
:158…160 °C
:Ambient
:201-771-8
- Appearance:White to off-white Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Loss on drying:≤1 %
- HPLC:≥97.5 % (ELSD)
- Specific optical rotation:-42° to -44° (20°C, 589 nm) (c=5, H2O)
Frequently Bought Together