Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Boron tribromide
Boron tribromide 1 mol/l in hexane, Sigma-Aldrich®
Boron tribromide 1 mol/l in hexane, Sigma-Aldrich®
SIAL211230-100ML
:
Merck
:
Boron tribromide 1 mol/l in hexane, Sigma-Aldrich®
SIAL211230-100ML
:
Merck
:
Pack type:
Glass bottle
Pk:
100 mL
Seq:
2440
:
:
250.52 g/mol
:
6.1,I
:
M-Clarity™ quality level = MQ200
:
233-657-9
:
3384
:
10294-33-4
:
BBr₃
:
MFCD00011312