5-Fluorocytosine
Catalog # PROOICRS0310
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:129.09 g/mol
Merck Index:13,04151
Boiling Pt:298 °C (1013 hPa)
Melting Pt:165 °C
EINECS:217-968-7
CAS Number:2022-85-7
Formula:C₄H₄FN₃O
MDL Number:MFCD00006035