Cytosine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 6-Aminopyrimidin-2(1H)-one, 4-Amino-2-pyrimidinol
CAS Number:71-30-7
:
Formula:C₄H₅N₃O
MDL Number:MFCD00006034
Density:0.99771 g/cm³ (25 °C)
Molecular Weight:111.1 g/mol
Melting Pt:>300 °C
Merck Index:13,02824
EINECS:200-749-5
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together