Order Entry
Poland
ContactUsLinkComponent
 
 
Diethylene glycol monomethyl ether, Sigma-Aldrich®
 :  Merck
 
 
Diethylene glycol monomethyl ether, Sigma-Aldrich®
 :  Merck
 

 

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -65 °C
  • Molar Mass:
    120.15 g/mol
  • Boiling Point:
    190 - 196 °C (1013 hPa)
  • Vapor Pressure:
    0.3 hPa (20 °C)
  • Flash Point:
    91 °C
  • Refractive Index:
    1.4264 (27 °C, 589 nm)
  • Explosion Limit:
    1.6 - 16.1 %(V)
  • Density:
    1.018 - 1.022 g/cm3 (20 °C) DIN 51757
  • pH:
    4 - 7 (200 g/l, H2O, 20 °C)
  • Ignition Point:
    215 °C
  • Evaporation number:
    900

 

Frequently Bought Together