Sodium metaarsenite, Sigma-Aldrich®
Catalog # SIALS7400-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: Sodium dioxoarsenate
:
:129.91 g/mol
EINECS:232-070-5
UN:2027
:GHS06
:GHS09
Storage Temperature:Ambient
CAS Number:7784-46-5
Formula:NaAsO₂
MDL Number:MFCD00003472
Melting Pt:615
Density:1.87 g/cm³ (20 °C)
ADR:6.1 II
Merck Index:13,08651