(Benzyl)triphenylphosphonium chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Benzyltriphenylphosphonium chloride
CAS Number:1100-88-5
:
Formula:C₂₅H₂₂ClP
MDL Number:MFCD00011913
Density:1.18 g/cm³ (20 °C)
Molecular Weight:388.88 g/mol
Melting Pt:345…347 °C
ADR:6.1 II
:M-Clarity™ quality level = MQ200
EINECS:214-154-3
UN:3464
:GHS06
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together