



Aniline hydrochloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:142-04-1
:
Formula:C₆H₈ClN
MDL Number:MFCD00012958
Merck Index:13,00616
Boiling Pt:245 °C (1013 hPa)
Melting Pt:198 °C
Density:1.2215 g/cm³ (20 °C)
Flash Pt:193 °C
EINECS:205-519-8
UN:1548
ADR:6.1,III
:GHS06
:GHS05
:GHS08
:GHS09
:129.59 g/mol
Frequently Bought Together