Tetramethylammonium hexafluorophosphate, Sigma-Aldrich®
Catalog # SIAL87725-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:219.11 g/mol
Melting Pt:300 °C
Density:1.617 g/cm³ (25 °C)
EINECS:209-194-3
UN:2923
ADR:8,III
CAS Number:558-32-7
Formula:C₄H₁₂F₆NP
MDL Number:MFCD00012012