Tetrakis(triphenylphosphine)palladium(0), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:14221-01-3
:
Formula:C₇₂H₆₀P₄Pd
MDL Number:MFCD00010012
:1155.58 g/mol
Melting Pt:103…107 °C
Density:1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:238-086-9
Frequently Bought Together