5-(4-Fluorophenyl)-1H-tetrazole, Sigma-Aldrich®
Catalog # SIAL681091-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:164.14 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:50907-21-6
Formula:C₇H₅FN₄
MDL Number:MFCD00478201