


N,N,N',N'-Tetramethyl-1,3-propanediamine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-95-2
:
Formula:C₇H₁₈N₂
MDL Number:MFCD00008337
:130.23 g/mol
Boiling Pt:145 °C (1013 hPa)
Density:0.78 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:203-818-8
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together