Copper(II) chloride dihydrate, Sigma-Aldrich®
SIAL459097-5G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
:
:170.48 g/mol
:13,02660
:100 °C
:2.54 g/cm³ (20 °C)
:Ambient
:231-210-2
:2802
:8,III
:M-Clarity™ quality level = MQ100
:GHS07
:GHS09
:10125-13-0
:CuCl₂·2H₂O
:MFCD00149674