Order Entry
Poland
ContactUsLinkComponent
Tris(4-fluorophenyl)phosphine, Sigma-Aldrich®
Tris(4-fluorophenyl)phosphine, Sigma-Aldrich®
Catalog # SIAL395099-5G
Supplier:  Merck
CAS Number:  
Tris(4-fluorophenyl)phosphine, Sigma-Aldrich®
Catalog # SIAL395099-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :316.26 g/mol
 :M-Clarity™ quality level = MQ200
CAS Number:18437-78-0
Formula:C₁₈H₁₂F₃P
MDL Number:MFCD00013553