(Ethyl)triphenylphosphonium iodide, Sigma-Aldrich®
Catalog # SIAL389471-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:418.26 g/mol
Melting Pt:163…166 °C
Density:1 g/cm³ (20 °C)
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:225-245-2
UN:2811
CAS Number:4736-60-1
Formula:C₂₀H₂₀IP
MDL Number:MFCD00040352