Order Entry
Poland
ContactUsLinkComponent
 
Irritant
Sodium deoxycholate monohydrate, Sigma-Aldrich®
Catalog # SIAL30968-25G
Supplier:  Merck
CAS Number:  
Irritant
Sodium deoxycholate monohydrate, Sigma-Aldrich®
Catalog # SIAL30968-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :432.58 g/mol
EINECS:206-132-7
 :GHS07
CAS Number:145224-92-6
Formula:C₂₄H₃₉NaO₄·1H₂O
MDL Number:MFCD00150750