Order Entry
Poland
ContactUsLinkComponent
Tris(4-chlorophenyl)phosphine, Sigma-Aldrich®
Tris(4-chlorophenyl)phosphine, Sigma-Aldrich®
Catalog # SIAL249491-1G
Supplier:  Merck
CAS Number:  
Tris(4-chlorophenyl)phosphine, Sigma-Aldrich®
Catalog # SIAL249491-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :365.63 g/mol
Boiling Pt:431 °C (1013 hPa)
Melting Pt:103 °C
Density:1.449 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:214-596-7
CAS Number:1159-54-2
Formula:C₁₈H₁₂Cl₃P
MDL Number:MFCD00013639