3-(Trimethylsilyl)-1-propanol, Sigma-Aldrich®
SIAL233803-5G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
:
:132.28 g/mol
:111 °C (1013 hPa)
:0.8324 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:2917-47-7
:HOH₂CCH₂CH₂Si(CH₃)₃